Bromoacetic acid N-hydroxysuccinimide ester, 90%
Bromoacetic acid N-hydroxysuccinimide ester, 90%
Bromoacetic acid N-hydroxysuccinimide ester, 90%
Thermo Scientific Chemicals

Bromoacetic acid N-hydroxysuccinimide ester, 90%

CAS: 42014-51-7 | C6H6BrNO4 | 236.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number 427780010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS42014-51-7
IUPAC Name2,5-dioxopyrrolidin-1-yl 2-bromoacetate
Molecular FormulaC6H6BrNO4
InChI KeyNKUZQMZWTZAPSN-UHFFFAOYSA-N
SMILESBrCC(=O)ON1C(=O)CCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to off-white
Elemental Analysis>=88 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only