7-Hydroxycoumarinyl-4-acetic acid, 97%
7-Hydroxycoumarinyl-4-acetic acid, 97%
7-Hydroxycoumarinyl-4-acetic acid, 97%
Thermo Scientific Chemicals

7-Hydroxycoumarinyl-4-acetic acid, 97%

CAS: 6950-82-9 | C11H8O5 | 220.18 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 427900050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6950-82-9
IUPAC Name2-(7-hydroxy-2-oxo-2H-chromen-4-yl)acetic acid
Molecular FormulaC11H8O5
InChI KeyBNHPMQBVNXMPDU-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC(=O)OC2=CC(O)=CC=C12
さらに表示
Appearance (Form)Crystalline powder
HPLC>=96.0 %
Appearance (Color)Off-white to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only