Tricyclohexylphosphine, 1M solution in toluene, AcroSeal™
Tricyclohexylphosphine, 1M solution in toluene, AcroSeal™
Tricyclohexylphosphine, 1M solution in toluene, AcroSeal™
Tricyclohexylphosphine, 1M solution in toluene, AcroSeal™
Thermo Scientific Chemicals

Tricyclohexylphosphine, 1M solution in toluene, AcroSeal™

CAS: 2622-14-2 | C18H33P | 280.43 g/mol
Have Questions?
Catalog NumberQuantityPackaging
428421000100 mLAcroSeal™ Glass Bottle
Catalog number 428421000
Price (USD)
-
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS2622-14-2
IUPAC Nametricyclohexylphosphane
Molecular FormulaC18H33P
InChI KeyWLPUWLXVBWGYMZ-UHFFFAOYSA-N
SMILESC1CCC(CC1)P(C1CCCCC1)C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Strength0.90 to 1.10 M
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only