(Trifluoromethyl)trimethylsilane, 0.5M solution in THF, AcroSeal™
(Trifluoromethyl)trimethylsilane, 0.5M solution in THF, AcroSeal™
(Trifluoromethyl)trimethylsilane, 0.5M solution in THF, AcroSeal™
(Trifluoromethyl)trimethylsilane, 0.5M solution in THF, AcroSeal™
Thermo Scientific Chemicals

(Trifluoromethyl)trimethylsilane, 0.5M solution in THF, AcroSeal™

CAS: 81290-20-2 | C5H11F3Si | 156.22 g/mol
Have Questions?
Catalog NumberQuantityPackaging
429171000100 mLAcroSeal™ Glass Bottle
Catalog number 429171000
Price (USD)
-
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS81290-20-2
IUPAC Nametert-butyl(trifluoromethyl)silane
Molecular FormulaC5H11F3Si
InChI KeyMTYSDPUZDZURPP-UHFFFAOYSA-N
SMILESCC(C)(C)[SiH2]C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Strength0.45 to 0.55 M
Appearance (Color)Clear colorless
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only