Methyl 2-aminopyridine-4-carboxylate, 98%
Methyl 2-aminopyridine-4-carboxylate, 98%
Methyl 2-aminopyridine-4-carboxylate, 98%
Thermo Scientific Chemicals

Methyl 2-aminopyridine-4-carboxylate, 98%

CAS: 6937-03-7 | C7H8N2O2 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 430060050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6937-03-7
IUPAC Namemethyl 2-aminopyridine-4-carboxylate
Molecular FormulaC7H8N2O2
InChI KeySVWWNEYBEFASMP-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=NC(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Infrared spectrumConforms
Melting point144°C to 148°C
Appearance (Color)White to brown
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only