4-Methylpyridine-3-boronic acid, 90%
4-Methylpyridine-3-boronic acid, 90%
4-Methylpyridine-3-boronic acid, 90%
Thermo Scientific Chemicals

4-Methylpyridine-3-boronic acid, 90%

CAS: 148546-82-1 | C6H8BNO2 | 136.95 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4302700101 gGlass bottle
Catalog number 430270010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Assay Percent Range88% min. (HPLC)
CAS Max %100.0
CAS Min %88.0
CAS148546-82-1
Chemical Name or Material4-Methylpyridine-3-boronic acid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only