1H-Benzimidazole-5-boronic acid pinacol ester, 97%
1H-Benzimidazole-5-boronic acid pinacol ester, 97%
1H-Benzimidazole-5-boronic acid pinacol ester, 97%
Thermo Scientific Chemicals

1H-Benzimidazole-5-boronic acid pinacol ester, 97%

CAS: 1007206-54-3 | C13H17BN2O2 | 244.10 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4305400101 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 430540010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1007206-54-3
IUPAC Name6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-1,3-benzodiazole
Molecular FormulaC13H17BN2O2
InChI KeyHCWNKNYTHLBIHX-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC=C2N=CNC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to tan to purple
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only