Ethyl chloroformate, 99%, AcroSeal™
Ethyl chloroformate, 99%, AcroSeal™
Ethyl chloroformate, 99%, AcroSeal™
Thermo Scientific Chemicals

Ethyl chloroformate, 99%, AcroSeal™

CAS: 541-41-3 | C3H5ClO2 | 108.52 g/mol
数量:
100 mL
800 mL
パッケージ:
すべて表示
AcroSeal™ Glass Bottle
製品番号(カタログ番号) 430798000
価格(JPY)
-
数量:
800 mL
パッケージ:
AcroSeal™ Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS541-41-3
IUPAC Nameethyl carbonochloridate
Molecular FormulaC3H5ClO2
InChI KeyRIFGWPKJUGCATF-UHFFFAOYSA-N
SMILESCCOC(Cl)=O
さらに表示
GC>=98.5 %
Appearance (Form)Clear liquid
Color scale=<50 APHA
Infrared spectrumConforms
GC=<0.3 % Ethanol
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only