Chlortetracycline hydrochloride, specified according to the requirements of Ph.Eur.
Chlortetracycline hydrochloride, specified according to the requirements of Ph.Eur.
Chlortetracycline hydrochloride, specified according to the requirements of Ph.Eur.
Thermo Scientific Chemicals

Chlortetracycline hydrochloride, specified according to the requirements of Ph.Eur.

Chlortetracycline hydrochloride, CAS # 64-72-2, is a broad-spectrum antibiotic of the tetracycline family. | CAS: 64-72-2 | C22H23ClN2O8·HCl | 515.33 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4309600505 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 430960050
Price (USD)
70.10
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
70.10
Each
Chemical Identifiers
CAS64-72-2
IUPAC Namehydrogen (2Z,4S,4aS,5aS,6S,12aS)-2-[amino(hydroxy)methylidene]-7-chloro-4-(dimethylamino)-6,10,11,12a-tetrahydroxy-6-methyl-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride
Molecular FormulaC22H24Cl2N2O8
InChI KeyRRXOSDMYCQSQHB-MRYPCDLHSA-N
SMILES[H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@H]3C(=C(O)C4=C(O)C=CC(Cl)=C4[C@@]3(C)O)C(=O)[C@]2(O)C(=O)C(=C(N)O)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Assay>=89.5 % Chlortetracycline hydrochloride,
Specific optical rotationOn dry substance
Water=<2.0 %
IdentificationPasses test
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General features
• Chlortetracycline hydrochloride is a broad-spectrum antibiotic of the tetracycline family
• Receives its name from a 7-chloro substitution on one of the rings of its parent structure, tetracycline
• A bacteriostatic that inhibits the 30S ribosomal subunit and prevents peptide formation
• Can work as an antiprotozoal and antifungal


Applications
• Chlortetracycline is suitable for use in biochemistry, microbiology, in vivo research, and cell culture
• Intended for research use only

RUO – Research Use Only

General References:

  1. Chen, C.; Ray, P.; Knowlton, K.F.; Pruden, A.; Xia, K. Effect of composting and soil type on dissipation of veterinary antibiotics in land-applied manures. Chemosphere. 2018, 196:270-279.
  2. Weinroth, M.D.; Scott, H.M.; Norby, B.; Loneragan, G.H.; Noyes, N.R.; Rovira, P.; Doster, E.; Yang, X.; Woerner, D.R.; Morley, P.S.; Belk, K.E. Effects of Ceftiofur and Chlortetracycline on the Resistomes of Feedlot Cattle. Appl Environ Microbiol. 2018, 84(13), e00610-18.