4-(2-Tetrahydro-2H-pyranoxy)phenylmagnesium bromide, 0.5M solution in THF, AcroSeal™
4-(2-Tetrahydro-2H-pyranoxy)phenylmagnesium bromide, 0.5M solution in THF, AcroSeal™
4-(2-Tetrahydro-2H-pyranoxy)phenylmagnesium bromide, 0.5M solution in THF, AcroSeal™
Thermo Scientific Chemicals

4-(2-Tetrahydro-2H-pyranoxy)phenylmagnesium bromide, 0.5M solution in THF, AcroSeal™

CAS: 36637-44-2 | C11H13BrMgO2 | 281.43 g/mol
Catalog NumberQuantityPackaging
43164050050 mLAcroSeal™ Glass Bottle
Catalog number 431640500
Price (JPY)
-
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS36637-44-2
IUPAC Name2-[4-(bromomagnesio)phenoxy]oxane
Molecular FormulaC11H13BrMgO2
InChI KeyOHPUQUBYKZJOLY-UHFFFAOYNA-M
SMILESBr[Mg]C1=CC=C(OC2CCCCO2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to brown
Appearance (Form)Solution
Titration Grignard0.40 to 0.60 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only