Imidazo[1,2-a]pyridine-2-carboxylic acid monohydrate, 97%
Imidazo[1,2-a]pyridine-2-carboxylic acid monohydrate, 97%
Imidazo[1,2-a]pyridine-2-carboxylic acid monohydrate, 97%
Thermo Scientific Chemicals

Imidazo[1,2-a]pyridine-2-carboxylic acid monohydrate, 97%

CAS: 64951-08-2 | C8H6N2O2 | 162.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 432780050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS64951-08-2
IUPAC Nameimidazo[1,2-a]pyridine-2-carboxylic acid
Molecular FormulaC8H6N2O2
InChI KeyWQLJLPDGSLZYEP-UHFFFAOYSA-N
SMILESOC(=O)C1=CN2C=CC=CC2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Additional infoHydrate, contains 9.5 % to 11.0 % water
Additional infoHydrate, contains 9.5 % to 11.0 % water
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only