Diphenyliodonium trifluoromethanesulfonate, 99%
Diphenyliodonium trifluoromethanesulfonate, 99%
Diphenyliodonium trifluoromethanesulfonate, 99%
Thermo Scientific Chemicals

Diphenyliodonium trifluoromethanesulfonate, 99%

CAS: 66003-76-7 | C12H10I·CF3O3S | 430.18 g/mol
Catalog NumberQuantity
4328400101 g
Catalog number 432840010
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS66003-76-7
IUPAC Namediphenyliodanium trifluoromethanesulfonate
Molecular FormulaC13H10F3IO3S
InChI KeySBQIJPBUMNWUKN-UHFFFAOYSA-M
SMILES[O-]S(=O)(=O)C(F)(F)F.[I+](C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
HPLC>=98.5 %
Melting point176°C to 180°C
Appearance (Color)White to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only