Tetrabutylammonium fluoride, 1M solution in THF, AcroSeal™
Tetrabutylammonium fluoride, 1M solution in THF, AcroSeal™
Tetrabutylammonium fluoride, 1M solution in THF, AcroSeal™
Tetrabutylammonium fluoride, 1M solution in THF, AcroSeal™
Thermo Scientific Chemicals

Tetrabutylammonium fluoride, 1M solution in THF, AcroSeal™

CAS: 429-41-4 | C16H36FN | 261.47 g/mol
数量:
100 mL
800 mL
パッケージ:
すべて表示
AcroSeal™ Glass bottle
製品番号(カタログ番号) 433518000
価格(JPY)
-
見積もりを依頼する
数量:
800 mL
パッケージ:
AcroSeal™ Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS429-41-4
IUPAC Nametetrabutylazanium fluoride
Molecular FormulaC16H36FN
InChI KeyFPGGTKZVZWFYPV-UHFFFAOYSA-M
SMILES[F-].CCCC[N+](CCCC)(CCCC)CCCC
さらに表示
Appearance (Form)Solution
Water=<8 %
Appearance (Color)Clear yellow to amber to light green to brown
Titration with HClO40.90 to 1.10 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only