N,N,N',N'-Tetramethylethylenediamine, 99.5%, purified by redistillation, AcroSeal™
N,N,N',N'-Tetramethylethylenediamine, 99.5%, purified by redistillation, AcroSeal™
N,N,N',N'-Tetramethylethylenediamine, 99.5%, purified by redistillation, AcroSeal™
N,N,N',N'-Tetramethylethylenediamine, 99.5%, purified by redistillation, AcroSeal™
Thermo Scientific Chemicals

N,N,N',N'-Tetramethylethylenediamine, 99.5%, purified by redistillation, AcroSeal™

CAS: 110-18-9 | C6H16N2 | 116.21 g/mol
Have Questions?
Catalog NumberQuantityPackaging
433831000100 mLAcroSeal™ Glass Bottle
Catalog number 433831000
Price (USD)
98.30
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
98.30
Each
Chemical Identifiers
CAS110-18-9
IUPAC Name[2-(dimethylamino)ethyl]dimethylamine
Molecular FormulaC6H16N2
InChI KeyKWYHDKDOAIKMQN-UHFFFAOYSA-N
SMILESCN(C)CCN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4170 to 1.4190 (20°C, 589 nm)
GC>=99.4 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only