Dilithium tetrabromonickelate(II), 0.5M solution in THF, AcroSeal™
Dilithium tetrabromonickelate(II), 0.5M solution in THF, AcroSeal™
Dilithium tetrabromonickelate(II), 0.5M solution in THF, AcroSeal™
Thermo Scientific Chemicals

Dilithium tetrabromonickelate(II), 0.5M solution in THF, AcroSeal™

CAS: 13826-95-4 | Br4Li2Ni | 392.19 g/mol
数量:
100 mL
800 mL
パッケージ:
AcroSeal™ Glass Bottle
製品番号(カタログ番号) 434548000
価格(JPY)
-
数量:
800 mL
パッケージ:
AcroSeal™ Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13826-95-4
Appearance (Form)Solution
Appearance (Color)Blue-green to brown
Titration Argentometric0.45 to 0.55 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only