N-Acetyl-D-galactosamine, 98%
N-Acetyl-D-galactosamine, 98%
N-Acetyl-D-galactosamine, 98%
Thermo Scientific Chemicals

N-Acetyl-D-galactosamine, 98%

CAS: 1811-31-0 | C8H15NO6 | 221.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 434920010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1811-31-0
IUPAC NameN-[(3R,4R,5R,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
Molecular FormulaC8H15NO6
InChI KeyOVRNDRQMDRJTHS-KEWYIRBNSA-N
SMILESCC(=O)N[C@H]1C(O)O[C@H](CO)[C@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=97.5 %
Specific optical rotation+80.0° to +90.0° (20°C, 589 nm) (c=1, water, after 5 hours)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only