(R)-(+)-2-(BOC-Amino)-1,4-butanediol, 97%, Thermo Scientific Chemicals
(R)-(+)-2-(BOC-Amino)-1,4-butanediol, 97%, Thermo Scientific Chemicals
(R)-(+)-2-(BOC-Amino)-1,4-butanediol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(R)-(+)-2-(BOC-Amino)-1,4-butanediol, 97%, Thermo Scientific Chemicals

CAS: 397246-14-9 | Molecular Formula: C9H19NO4 | Molecular Weight: 205.25 g/mol
Have Questions?
Catalog NumberQuantity
4355700505 g
Catalog number 435570050
Price (TWD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS397246-14-9
IUPAC Nametert-butyl N-(1,4-dihydroxybutan-2-yl)carbamate
Molecular FormulaC9H19NO4
InChI KeyKLRRFBSWOIUAHZ-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)NC(CO)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
GC>=96.0 %
Appearance (Color)White
Melting point65°C to 69°C
Specific optical rotation+7° to +9° (20°C, 589 nm) (c=1, chloroform)
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only