1-Methyl-1-propenylmagnesium bromide, 0.5M solution in THF, AcroSeal™
1-Methyl-1-propenylmagnesium bromide, 0.5M solution in THF, AcroSeal™
1-Methyl-1-propenylmagnesium bromide, 0.5M solution in THF, AcroSeal™
Thermo Scientific Chemicals

1-Methyl-1-propenylmagnesium bromide, 0.5M solution in THF, AcroSeal™

CAS: 85676-85-3 | C4H7BrMg | 159.31 g/mol
数量:
100 mL
800 mL
パッケージ:
AcroSeal™ Glass Bottle
製品番号(カタログ番号) 436028000
価格(JPY)
-
数量:
800 mL
パッケージ:
AcroSeal™ Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS85676-85-3
IUPAC Namemagnesium(2+) but-2-en-2-ide bromide
Molecular FormulaC4H7BrMg
InChI KeyFGYXQBPXHQHYNU-UHFFFAOYSA-M
SMILES[Mg++].[Br-].CC=[C-]C
さらに表示
Titration Grignard0.45 to 0.55 M
Appearance (Color)Colorless to dark yellow
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only