Thermo Scientific Chemicals

N-(tert-Butoxycarbonyloxy)succinimide, 97%, Thermo Scientific Chemicals

Catalog number: 436240250
25 g, Each
Thermo Scientific Chemicals

N-(tert-Butoxycarbonyloxy)succinimide, 97%, Thermo Scientific Chemicals

Catalog number: 436240250
25 g, Each
Quantity

Chemical Identifiers

CAS
13139-12-3
IUPAC Name
tert-butyl 2,5-dioxopyrrolidin-1-yl carbonate
Molecular Formula
C9H13NO5
InChI Key
VTGFSVGZCYYHLO-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)ON1C(=O)CCC1=O
Appearance (Color)
White
Appearance (Form)
Powder or solid
Infrared spectrum
Conforms
Elemental Analysis
48.22 to 52.24 % C
Elemental Analysis
6.25 to 6.77 % N

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text