(S)-3-(BOC-amino)piperidina, 97 %, Thermo Scientific Chemicals
(S)-3-(BOC-amino)piperidina, 97 %, Thermo Scientific Chemicals
(S)-3-(BOC-amino)piperidina, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(S)-3-(BOC-amino)piperidina, 97 %, Thermo Scientific Chemicals

CAS: 216854-23-8 | C10H20N2O2 | 200.28 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
Número de catálogo 436530050
Precio (USD)
-
Cantidad:
5 g
Pedido a granel o personalizado
Identificadores químicos
CAS216854-23-8
IUPAC Nametert-butyl N-(piperidin-3-yl)carbamate
Molecular FormulaC10H20N2O2
InChI KeyWUOQXNWMYLFAHT-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)NC1CCCNC1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
GC>=96.0 %
Infrared spectrumConforms
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Specific optical rotation-2.5° to -3.5° (20°C, 589 nm) (c=1 in DMF)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only