1-BOC-3-Aminopiperidine, 97%
1-BOC-3-Aminopiperidine, 97%
1-BOC-3-Aminopiperidine, 97%
Thermo Scientific Chemicals

1-BOC-3-Aminopiperidine, 97%

CAS: 144243-24-3 | C10H20N2O2 | 200.28 g/mol
Have Questions?
Catalog NumberQuantity
4366000505 g
Catalog number 436600050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS144243-24-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only