(R)-(-)-tert-Leucinol, 98%
(R)-(-)-tert-Leucinol, 98%
(R)-(-)-tert-Leucinol, 98%
Thermo Scientific Chemicals

(R)-(-)-tert-Leucinol, 98%

CAS: 112245-09-7 | C6H15NO | 117.19 g/mol
Have Questions?
Catalog NumberQuantity
436671000100 mg
Catalog number 436671000
Price (USD)
-
Quantity:
100 mg
Request bulk or custom format
Chemical Identifiers
CAS112245-09-7
IUPAC Name(2R)-2-amino-3,3-dimethylbutan-1-ol
Molecular FormulaC6H15NO
InChI KeyJBULSURVMXPBNA-MLHKIVSYNA-N
SMILESCC(C)(C)[C@@H](N)CO
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Form)Liquid or low melting solid
Specific optical rotation-34° to -40° (20°C, 589 nm) (c=0.7, ethanol)
Appearance (Color)Colorless to yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only