N,N,N',N'-Tetramethylchloroformamidinium hexafluorophosphat
Thermo Scientific Chemicals

N,N,N',N'-Tetramethylchloroformamidinium hexafluorophosphat

CAS: 94790-35-9 | C5H12ClN2 | 135.61 g/mol
製品番号(カタログ番号)数量
43907025025 g
製品番号(カタログ番号) 439070250
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS94790-35-9
IUPAC Name[bis(dimethylamino)methylidene]-λ³-chloranylium
Molecular FormulaC5H12ClN2
InChI KeyQZFWNEASXBNLCD-UHFFFAOYSA-N
SMILESCN(C)C(=[Cl+])N(C)C
さらに表示
HPLC>=97.5 %
Appearance (Color)White to almost white
Infrared spectrumConforms
Melting point100°C to 106°C
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only