(±)-BOC-alpha-Phosphonoglycine trimethyl ester, 97%
(±)-BOC-alpha-Phosphonoglycine trimethyl ester, 97%
(±)-BOC-alpha-Phosphonoglycine trimethyl ester, 97%
Thermo Scientific Chemicals

(±)-BOC-alpha-Phosphonoglycine trimethyl ester, 97%

CAS: 89524-98-1 | C10H20NO7P | 297.24 g/mol
Have Questions?
Catalog NumberQuantity
4392100505 g
Catalog number 439210050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS89524-98-1
IUPAC Namemethyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-2-(dimethoxyphosphoryl)acetate
Molecular FormulaC10H20NO7P
InChI KeyLJHAPRKTPAREGO-ZETCQYMHSA-N
SMILESCOC(=O)[C@@H](NC(=O)OC(C)(C)C)P(=O)(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
GC>=96.0 %
Infrared spectrumConforms
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only