N-BOC-1-Aminocyclobutanecarboxylic acid, 97%, Thermo Scientific Chemicals
N-BOC-1-Aminocyclobutanecarboxylic acid, 97%, Thermo Scientific Chemicals
N-BOC-1-Aminocyclobutanecarboxylic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-BOC-1-Aminocyclobutanecarboxylic acid, 97%, Thermo Scientific Chemicals

CAS: 120728-10-1 | Molecular Formula: C10H16NO4 | Molecular Weight: 214.24 g/mol
Have Questions?
Catalog NumberQuantity
4404400505 g
Catalog number 440440050
Price (USD)
347.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
347.00
Each
Chemical Identifiers
CAS120728-10-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Appearance (Form)Powder
Infrared spectrumConforms
Titration with HClO496.0 to 104.0 %

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only