4-Pyridinepropanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Pyridinepropanol, Thermo Scientific Chemicals

製品番号(カタログ番号) 440560250
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2629-72-3
IUPAC Name3-(pyridin-4-yl)propan-1-ol
Molecular FormulaC8H11NO
InChI KeyPZVZGDBCMQBRMA-UHFFFAOYSA-N
SMILESOCCCC1=CC=NC=C1
さらに表示
Appearance (Form)Powder and/or lumps
GC>=96.0 %
Infrared spectrumConforms
Melting point35°C to 39°C
Appearance (Color)White to grey to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only