(7-Azabenzotriazol-1-yloxy)tripyrrolidinophosphonium hexafluorophosphate, 98%
(7-Azabenzotriazol-1-yloxy)tripyrrolidinophosphonium hexafluorophosphate, 98%
(7-Azabenzotriazol-1-yloxy)tripyrrolidinophosphonium hexafluorophosphate, 98%
Thermo Scientific Chemicals

(7-Azabenzotriazol-1-yloxy)tripyrrolidinophosphonium hexafluorophosphate, 98%

CAS: 156311-83-0 | C17H27F6N7OP2 | 521.39 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4406100505 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 440610050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS156311-83-0
IUPAC Namehexafluoro-λ⁵-phosphanuide; tris(pyrrolidin-1-yl)({3H-[1,2,3]triazolo[4,5-b]pyridin-3-yloxy})phosphanium
Molecular FormulaC17H27F6N7OP2
InChI KeyCBZAHNDHLWAZQC-UHFFFAOYSA-N
SMILESF[P-](F)(F)(F)(F)F.C1CCN(C1)[P+](ON1N=NC2=CC=CN=C12)(N1CCCC1)N1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
Elemental Analysis38.18 to 40.14 % (C)
Elemental Analysis18.33 to 19.28 % (N)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only