96-Well Septa Dx
Applied Biosystems™

96-Well Septa Dx

This is a 96-well septa designed for use with Applied Biosystems Dx genetic analyzers.
Have Questions?
Catalog NumberQuantity
441070020 Pk.
Catalog number 4410700
Price (USD)
689.65
Online Exclusive
734.00
Save 44.35 (6%)
Each
Quantity:
20 Pk.
Price (USD)
689.65
Online Exclusive
734.00
Save 44.35 (6%)
Each

This is a 96-well septa designed for use with Applied Biosystems Dx genetic analyzers.

Note: Applied Biosystems Dx genetic analyzers and consumables are for distribution and use in select countries only.

This is a 96-well septa designed for use with the 3500 Dx/3500xL Dx Genetic Analyzer.

Septa and plate base/retainer kits for 8 tube strip, 96 and 384-well plates used with the 3500 Dx Series Genetic Analyzers.

Note: The 3500 Dx Series Systems and consumables are for distribution and use in select countries only.

For In Vitro Diagnostic Use Only
Specifications
Compatibility96-Well Microtiter Plates, MicroAmp™ Optical 96-Well Reaction Plates
For Use With (Equipment)3500 xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500 xL Genetic Analyzer, 3500 Genetic Analyzer
Format96-well
Product TypeSepta
Quantity20 Pk.
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Ambient storage.
Each Pk. contains 20 sets.