D-Cysteine hydrochloride monohydrate, 98%
D-Cysteine hydrochloride monohydrate, 98%
D-Cysteine hydrochloride monohydrate, 98%
Thermo Scientific Chemicals

D-Cysteine hydrochloride monohydrate, 98%

CAS: 207121-46-8 | C3H10ClNO3S | 175.627 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
500 mg
1 g
Packaging:
Glass Bottle
Catalog number 442780010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS207121-46-8
IUPAC Name2-amino-3-sulfanylpropanoic acid hydrate hydrochloride
Molecular FormulaC3H10ClNO3S
InChI KeyQIJRTFXNRTXDIP-UHFFFAOYNA-N
SMILESO.Cl.NC(CS)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Residue after ignition=<0.2 %
Infrared spectrumConforms
Specific optical rotation-5.0° to -7.0° (20°C, 589 nm) (c=8, 6M HCl)
Loss on drying9.99 % to 10.45 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only