Methyl 4-aminobutyrate hydrochloride, 99%
Methyl 4-aminobutyrate hydrochloride, 99%
Methyl 4-aminobutyrate hydrochloride, 99%
Thermo Scientific Chemicals

Methyl 4-aminobutyrate hydrochloride, 99%

CAS: 13031-60-2 | C5H12ClNO2 | 153.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass Bottle
Catalog number 442910250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS13031-60-2
IUPAC Name4-methoxy-4-oxobutan-1-aminium chloride
Molecular FormulaC5H12ClNO2
InChI KeyWPGPRLVPWACBHW-UHFFFAOYSA-N
SMILES[Cl-].COC(=O)CCC[NH3+]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric98.5 to 101.5 %
Infrared spectrumConforms
Appearance (Color)White to off-white
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only