BOC-N-Methyl-L-alanine, 99%
BOC-N-Methyl-L-alanine, 99%
BOC-N-Methyl-L-alanine, 99%
Thermo Scientific Chemicals

BOC-N-Methyl-L-alanine, 99%

CAS: 16948-16-6 | C9H17NO4 | 203.24 g/mol
製品番号(カタログ番号) 442930010
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS16948-16-6
IUPAC Name(2S)-2-{[(tert-butoxy)carbonyl](methyl)amino}propanoic acid
Molecular FormulaC9H17NO4
InChI KeyVLHQXRIIQSTJCQ-LURJTMIESA-N
SMILESC[C@H](N(C)C(=O)OC(C)(C)C)C(O)=O
さらに表示
Infrared spectrumConforms
Appearance (Color)White to off-white
Titration with NaOH98.5 to 101.5 %
Appearance (Form)Powder
Specific optical rotation-29° to -31° (20°C, 589 nm) (c=1 in EtOH)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only