Vinylmagnesium bromide, 1M solution in 2-MeTHF, AcroSeal™
Vinylmagnesium bromide, 1M solution in 2-MeTHF, AcroSeal™
Vinylmagnesium bromide, 1M solution in 2-MeTHF, AcroSeal™
Thermo Scientific Chemicals

Vinylmagnesium bromide, 1M solution in 2-MeTHF, AcroSeal™

CAS: 1826-67-1 | C2H3BrMg | 131.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Catalog number 445971000
Price (USD)
112.65
Special offer
Online exclusive
Ends: 28-Dec-2026
132.00
Save 19.35
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
112.65
Special offer
Online exclusive
Ends: 28-Dec-2026
132.00
Save 19.35
Each
Chemical Identifiers
CAS1826-67-1
IUPAC Namebromo(ethenyl)magnesium
Molecular FormulaC2H3BrMg
InChI KeyXHHHAXOHMKAOSL-UHFFFAOYSA-M
SMILESBr[Mg]C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear to hazy, yellow to brown to black
Appearance (Form)Solution
AppearanceMay develop some turbidity or precipitate
Titration Active base0.80 to 1.10 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only