2-Iodosobenzoic acid, 98%
2-Iodosobenzoic acid, 98%
2-Iodosobenzoic acid, 98%
Thermo Scientific Chemicals

2-Iodosobenzoic acid, 98%

CAS: 304-91-6 | C7H5IO3 | 264.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 446030010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS304-91-6
IUPAC Name2-iodosylbenzoic acid
Molecular FormulaC7H5IO3
InChI KeyIFPHDUVGLXEIOQ-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1[I]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
HPLC>=97.5 %
Infrared spectrumConforms
Titration Iodometric97.5 to 102.5 %
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
oxidation reagent
RUO – Research Use Only