Calcium phosphate, dibasic, 98+%, for analysis, anhydrous
Calcium phosphate, dibasic, 98+%, for analysis, anhydrous
Calcium phosphate, dibasic, 98+%, for analysis, anhydrous
Thermo Scientific Chemicals

Calcium phosphate, dibasic, 98+%, for analysis, anhydrous

CAS: 7757-93-9 | CaHO4P | 136.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Plastic bottle
Catalog number 446432500
Price (TWD)
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7757-93-9
IUPAC Namecalcium hydrogen phosphate
Molecular FormulaCaHO4P
InChI KeyFUFJGUQYACFECW-UHFFFAOYSA-L
SMILES[Ca++].OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Loss on ignition6.6 % to 8.5 %
Arsenic (As)=<0.0003 %
Appearance (Form)Powder
Titration Complexometric98.0 to 102.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only