4-(Trifluoromethyl)-1H-pyrazole, 97%
4-(Trifluoromethyl)-1H-pyrazole, 97%
4-(Trifluoromethyl)-1H-pyrazole, 97%
Thermo Scientific Chemicals

4-(Trifluoromethyl)-1H-pyrazole, 97%

CAS: 52222-73-8 | C4H3F3N2 | 136.08 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4464900101 gGlass bottle
Catalog number 446490010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS52222-73-8
IUPAC Name4-(trifluoromethyl)-1H-pyrazole
Molecular FormulaC4H3F3N2
InChI KeyKDEJQUNODYXYBJ-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CNN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Crystalline solid
Infrared spectrumConforms
GC>=96 %
Water=<2.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only