Sodium phosphate, dibasic dodecahydrate, 99%, for biochemistry
Sodium phosphate, dibasic dodecahydrate, 99%, for biochemistry
Sodium phosphate, dibasic dodecahydrate, 99%, for biochemistry
Thermo Scientific Chemicals

Sodium phosphate, dibasic dodecahydrate, 99%, for biochemistry

CAS: 10039-32-4 ∣ HNa2O4P·12H2O ∣ 358.13 g/mol
製品番号(カタログ番号)数量
4472500252.5 kg
製品番号(カタログ番号) 447250025
価格(JPY)
-
数量:
2.5 kg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS10039-32-4
IUPAC Namedisodium dodecahydrate hydrogen phosphate
Molecular FormulaH25Na2O16P
InChI KeyDGLRDKLJZLEJCY-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].OP([O-])([O-])=O
さらに表示
Potassium (K)=<100 ppm
Appearance (Color)White
SolubilityClear colorless
UVat 260 nm A: =<0.01
UV(0.1 M in water)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only