Crystal violet, ACS reagent
Crystal violet, ACS reagent
Crystal violet, ACS reagent
Thermo Scientific Chemicals

Crystal violet, ACS reagent

CAS: 548-62-9 | C25H30ClN3 | 407.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
View all
Glass bottle
Catalog number 447572500
Price (USD)
223.00
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
223.00
Each
Chemical Identifiers
CAS548-62-9
IUPAC Nametris[4-(dimethylamino)phenyl]methylium chloride
Molecular FormulaC25H30ClN3
InChI KeyZXJXZNDDNMQXFV-UHFFFAOYSA-M
SMILES[Cl-].CN(C)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay>=90.0 % (on dry substance)
Loss on drying=<7.5 %
Absorbance characteristics(0.98-1.9) Passes test
SensitivityPasses test
Appearance (Color)Dark green
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only