Bisulfato de potasio, 99 %, extra puro, Thermo Scientific Chemicals
Bisulfato de potasio, 99 %, extra puro, Thermo Scientific Chemicals
Bisulfato de potasio, 99 %, extra puro, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bisulfato de potasio, 99 %, extra puro, Thermo Scientific Chemicals

Bisulfato de potasio, 99 %, HKO4S, número de CAS 7646-93-7, bisulfato potásico, ácido sulfúrico, sal monopotásica, sal enixum, hidrogensulfato de potasio, sulfato de potasio ácido, sulfato de ácido potásico, sulfato de hidrógeno de potasio, sulfato monopotásico, Caswell n.º 682c, 500 g
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
500 g
2.5 kg
10 kg
Envase:
Ver todo
Frasco de vidrio
Número de catálogo 447615000
Precio (USD)
-
Cantidad:
500 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS7646-93-7
IUPAC Namepotassium hydrogen sulfate
Molecular FormulaHKO4S
InChI KeyCHKVPAROMQMJNQ-UHFFFAOYSA-M
SMILES[K+].OS([O-])(=O)=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Ammonium (NH4)=<0.003 %
Appearance (Form)Crystalline powder
Titration with NaOH>=98.5 %
Solubility(10 % in water) Clear colorless
Chloride (Cl)=<0.002 %
Ver más
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only