Lead(II) perchlorate trihydrate, ACS reagent
Lead(II) perchlorate trihydrate, ACS reagent
Lead(II) perchlorate trihydrate, ACS reagent
Thermo Scientific Chemicals

Lead(II) perchlorate trihydrate, ACS reagent

CAS: 13453-62-8 | Cl2H6O11Pb | 460.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 447781000
Price (USD)
74.65
Special offer
Online exclusive
Ends: 28-Dec-2026
87.80
Save 13.15
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
74.65
Special offer
Online exclusive
Ends: 28-Dec-2026
87.80
Save 13.15
Each
Chemical Identifiers
CAS13453-62-8
IUPAC Nameλ²-lead(2+) trihydrate diperchlorate
Molecular FormulaCl2H6O11Pb
InChI KeyKKGGEAQRICYXNM-UHFFFAOYSA-L
SMILESO.O.O.[Pb++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Titration Complexometric97.0 to 102.0 %
pH3.0 to 5.0 (5% in water, 25.0°C)
Insoluble matter=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only