Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]dichloropalladium, 95%
Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]dichloropalladium, 95%
Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]dichloropalladium, 95%
Thermo Scientific Chemicals

Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]dichloropalladium, 95%

CAS: 887919-35-9 | C32H56Cl2N2P2Pd | 708.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 447890010
Price (USD)
182.65
Special offer
Online exclusive
Ends: 28-Dec-2026
215.00
Save 32.35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
182.65
Special offer
Online exclusive
Ends: 28-Dec-2026
215.00
Save 32.35
Each
Chemical Identifiers
CAS887919-35-9
IUPAC Namebis(4-(di-tert-butylphosphanyl)-N,N-dimethylaniline); dichloropalladium
Molecular FormulaC32H56Cl2N2P2Pd
InChI KeyDWOZNANUEDYIOF-UHFFFAOYSA-L
SMILESCl[Pd]Cl.CN(C)C1=CC=C(C=C1)P(C(C)(C)C)C(C)(C)C.CN(C)C1=CC=C(C=C1)P(C(C)(C)C)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder and/or chunks
Infrared spectrumConforms
HPLC>=94 %
Appearance (Color)Yellow to dark yellow to dark orange
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only