Chloroacetic acid, 99+%, ACS reagent
Chloroacetic acid, 99+%, ACS reagent
Chloroacetic acid, 99+%, ACS reagent
Thermo Scientific Chemicals

Chloroacetic acid, 99+%, ACS reagent

CAS: 79-11-8 | C2H3ClO2 | 94.5 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 448111000
Price (USD)
61.65
Online Exclusive
68.20
Save 6.55 (10%)
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
61.65
Online Exclusive
68.20
Save 6.55 (10%)
Each
Chemical Identifiers
CAS79-11-8
IUPAC Name2-chloroacetic acid
Molecular FormulaC2H3ClO2
InChI KeyFOCAUTSVDIKZOP-UHFFFAOYSA-N
SMILESOC(=O)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=99.0 %
Chloride (Cl)=<0.01 %
Sulfate (SO4)=<0.02 %
Appearance (Color)Colorless to white
Sulfated ash=<0.02 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only