L(+)-Ascorbyl palmitate, specified according to requirements of USP/Ph.Eur., Thermo Scientific Chemicals
L(+)-Ascorbyl palmitate, specified according to requirements of USP/Ph.Eur., Thermo Scientific Chemicals
L(+)-Ascorbyl palmitate, specified according to requirements of USP/Ph.Eur., Thermo Scientific Chemicals
Thermo Scientific Chemicals

L(+)-Ascorbyl palmitate, specified according to requirements of USP/Ph.Eur., Thermo Scientific Chemicals

Have Questions?
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 449480250
Price (USD)
61.80
Each
Request bulk or custom format
Price (USD)
61.80
Each
Chemical Identifiers
CAS137-66-6
IUPAC Name2-(4,5-dihydroxy-3-oxo-2,3-dihydrofuran-2-yl)-2-hydroxyethyl hexadecanoate
Molecular FormulaC22H38O7
InChI KeyNULLRXHRYOXSEW-UHFFFAOYNA-N
SMILESCCCCCCCCCCCCCCCC(=O)OCC(O)C1OC(O)=C(O)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Titration Iodimetric98.0 to 100.5 % (on dry substance)
Specific optical rotation+21° to +24° (20°C, 589 nm) (c=10, methanol)
Appearance (Form)Powder
Melting point107°C to 117°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only