1-(1-Naphthyl)ethylamine, 98%
1-(1-Naphthyl)ethylamine, 98%
1-(1-Naphthyl)ethylamine, 98%
Thermo Scientific Chemicals

1-(1-Naphthyl)ethylamine, 98%

CAS: 42882-31-5 | C12H13N | 171.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Catalog number 450570050
Price (EUR)
50,40
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
50,40
Each
Chemical Identifiers
CAS42882-31-5
IUPAC Name1-(naphthalen-1-yl)ethan-1-amine
Molecular FormulaC12H13N
InChI KeyRTCUCQWIICFPOD-UHFFFAOYNA-N
SMILESCC(N)C1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.6220 to 1.6240 (20°C, 589 nm)
Infrared spectrumConforms
Titration with HClO4>=97.5 %
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only