Sodium hydroxide, 98%, pure, flakes
Sodium hydroxide, 98%, pure, flakes
Sodium hydroxide, 98%, pure, flakes
Thermo Scientific Chemicals

Sodium hydroxide, 98%, pure, flakes

CAS: 1310-73-2 | HNaO | 39.997 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
10 kg
Catalog number 450580100
Price (USD)
192.65
Online exclusive
214.00
Save 21.35
Each
Quantity:
10 kg
Request bulk or custom format
Price (USD)
192.65
Online exclusive
214.00
Save 21.35
Each
Chemical Identifiers
CAS1310-73-2
IUPAC Namesodium hydroxide
Molecular FormulaHNaO
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES[OH-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Flakes
Appearance (Color)White
Titration with HCl>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only