3-Butenylamine, 97%
3-Butenylamine, 97%
3-Butenylamine, 97%
Thermo Scientific Chemicals

3-Butenylamine, 97%

CAS: 2524-49-4 | C4H9N | 71.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 455050050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS2524-49-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.4215 to 1.4275 (20°C, 589 nm)
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only