Thermo Scientific Chemicals

Amodiaquine dihydrochloride dihydrate

Catalog number: 455200250
25g, Each
Thermo Scientific Chemicals

Amodiaquine dihydrochloride dihydrate

Catalog number: 455200250
25g, Each
Quantity

Chemical Identifiers

CAS
6398-98-7
IUPAC Name
dihydrogen 4-[(7-chloroquinolin-4-yl)amino]-2-[(diethylamino)methyl]phenol dihydrate dichloride
Molecular Formula
C20H28Cl3N3O3
InChI Key
YVNAYSHNIILOJS-UHFFFAOYSA-N
SMILES
[H+].[H+].O.O.[Cl-].[Cl-].CCN(CC)CC1=CC(NC2=C3C=CC(Cl)=CC3=NC=C2)=CC=C1O
Appearance (Color)
Yellow
Infrared spectrum
Conforms
Water
7.0 to 9.0 %
Appearance (Form)
Crystalline powder
Melting point
164°C to 169°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text