Nystatin, 85+%
Nystatin, 85+%
Nystatin, 85+%
Thermo Scientific Chemicals

Nystatin, 85+%

製品番号(カタログ番号)数量
45550025025 g
製品番号(カタログ番号) 455500250
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1400-61-9
IUPAC Name(1S,15S,16R,17R,18S,19E,21E,25E,27E,29E,31E)-33-{[(2S,3S,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy}-1,3,4,7,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,25,27,29,31-hexaene-36-carboxylic acid
Molecular FormulaC47H75NO17
InChI KeyVQOXZBDYSJBXMA-RKEBNKJGSA-N
SMILESC[C@H]1O[C@H](OC2CC3O[C@@](O)(CC(O)C3C(O)=O)CC(O)C(O)CCC(O)CC(O)CC(O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)C=CC=CCCC=CC=CC=CC=C2)[C@@H](O)[C@@H](N)[C@@H]1O
さらに表示
Appearance (Form)Powder
HPLC>=85.0 %
UVat 305 nm A: >=0.60
Activity>=4400 IU/mg (dried substance)
Water=<6 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only