N,N-Diethylpropargylamine, 97%
N,N-Diethylpropargylamine, 97%
N,N-Diethylpropargylamine, 97%
Thermo Scientific Chemicals

N,N-Diethylpropargylamine, 97%

CAS: 4079-68-9 | C7H13N | 111.18 g/mol
Quantity:
5 g
25 g
100 g
Catalog number 455580250
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS4079-68-9
IUPAC Namediethyl(prop-2-yn-1-yl)amine
Molecular FormulaC7H13N
InChI KeyJZJXKEWVUBVOEH-UHFFFAOYSA-N
SMILESCCN(CC)CC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only