2-Methyl-4-nitrobenzoic acid, 98%
2-Methyl-4-nitrobenzoic acid, 98%
2-Methyl-4-nitrobenzoic acid, 98%
Thermo Scientific Chemicals

2-Methyl-4-nitrobenzoic acid, 98%

CAS: 1975-51-5 | C8H7NO4 | 181.15 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) 457811000
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1975-51-5
IUPAC Name2-methyl-4-nitrobenzoic acid
Molecular FormulaC8H7NO4
InChI KeyXXXOBNJIIZQSPT-UHFFFAOYSA-N
SMILESCC1=CC(=CC=C1C(O)=O)[N+]([O-])=O
さらに表示
Appearance (Color)White to yellow
Infrared spectrumConforms
Appearance (Form)Powder
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only