Ethylenediaminetetraacetic acid disodium magnesium salt hydrate, 97%
Ethylenediaminetetraacetic acid disodium magnesium salt hydrate, 97%
Ethylenediaminetetraacetic acid disodium magnesium salt hydrate, 97%
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid disodium magnesium salt hydrate, 97%

CAS: 194491-32-2 | C10H12MgN2Na2O8 | 358.50 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 457931000
Price (USD)
40.65
Special offer
Online exclusive
Ends: 28-Dec-2026
48.00
Save 7.35
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
40.65
Special offer
Online exclusive
Ends: 28-Dec-2026
48.00
Save 7.35
Each
Chemical Identifiers
CAS194491-32-2
IUPAC Namemagnesium(2+) disodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
Molecular FormulaC10H12MgN2Na2O8
InChI KeyAWNVVAMWLMUZOZ-UHFFFAOYSA-J
SMILES[Na+].[Na+].[Mg++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Powder or crystalline powder or crystals
Infrared spectrumConforms
Loss on drying10 % to 20 % (200°C) (constant wt.)
Assay>=96.0 % (On dry substance)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only